Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334459.1 | NP_001320784.1 | 4 - 5 | 938 | 82 | |
NM_105719.5 | NP_177208.2 | 4 - 5 | 926 | 82 | ||
GenBank mRNA | U14175.1 | - | 4 - 5 | 837 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |