Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATWHY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105795.5 | NP_177282.2 | 7 - 8 | 648 | 76 | |
GenBank mRNA | AB493532.1 | - | 7 - 8 | 585 | 76 | |
AY072110.1 | - | 7 - 8 | 612 | 76 | ||
AY122961.1 | - | 7 - 8 | 585 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |