Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105813.4 | NP_565017.1 | 6 - 7 | 917 | 124 | |
GenBank mRNA | AK118835.1 | - | 6 - 7 | 870 | 124 | |
BT002403.1 | - | 6 - 7 | 872 | 124 | ||
BT008888.1 | - | 6 - 7 | 859 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |