Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105827.3 | NP_177314.2 | 6 - 7 | 949 | 105 | |
NM_202397.2 | NP_974126.1 | 7 - 8 | 807 | 105 | ||
GenBank mRNA | AJ555410.1 | - | 7 - 8 | 686 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |