Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198444.1 | NP_001185373.1 | 2 - 3 | 590 | 67 | |
NM_105835.2 | NP_177322.2 | 2 - 3 | 608 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |