Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G71810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105839.3 | NP_565025.1 | 11 - 12 | 1459 | 70 | |
GenBank mRNA | AY039896.1 | - | 11 - 12 | 1362 | 70 | |
AY094006.1 | - | 11 - 12 | 1239 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |