Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 12 - 13 | 1419 | 105 | |
NM_105840.4 | NP_565026.1 | 12 - 13 | 1272 | 105 | ||
GenBank mRNA | AY046014.1 | - | 12 - 13 | 1193 | 105 | |
AY133819.1 | - | 11 - 12 | 924 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |