Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334523.1 | NP_001321803.1 | 4 - 5 | 1244 | 119 | |
NM_105891.3 | NP_177376.1 | 4 - 5 | 1094 | 119 | ||
NM_179547.5 | NP_849878.2 | 4 - 5 | 1029 | 119 | ||
NM_202401.1 | NP_974130.1 | 4 - 5 | 999 | 119 | ||
GenBank mRNA | AY035041.1 | - | 4 - 5 | 1036 | 119 | |
AY051073.1 | - | 3 - 4 | 925 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |