Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105930.3 | NP_177415.1 | 1 - 2 | 437 | 146 | |
GenBank mRNA | AY059850.1 | - | 1 - 2 | 363 | 146 | |
AY943843.1 | - | 1 - 2 | 81 | 146 | ||
BT010328.1 | - | 1 - 2 | 81 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |