Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIP3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105978.4 | NP_177462.1 | 1 - 2 | 353 | 63 | |
GenBank mRNA | AY064054.1 | - | 1 - 2 | 176 | 63 | |
AY117339.1 | - | 1 - 2 | 142 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |