Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334584.1 | NP_001322854.1 | 8 - 9 | 884 | 98 | |
NM_001334585.1 | NP_001322853.1 | 7 - 8 | 760 | 98 | ||
NM_105990.2 | NP_177474.1 | 8 - 9 | 884 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |