Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344133.1 | NP_001331884.1 | 1 - 2 | 1742 | 106 | |
NM_123013.4 | NP_568533.2 | 2 - 3 | 1163 | 106 | ||
GenBank mRNA | AY093766.1 | - | 2 - 3 | 911 | 106 | |
BT001045.1 | - | 2 - 3 | 894 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |