Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344150.1 | NP_001329981.1 | 18 - 19 | 2081 | 94 | |
NM_001344151.1 | NP_001329982.1 | 18 - 19 | 2081 | 94 | ||
NM_001344152.1 | NP_001329979.1 | 17 - 18 | 2152 | 94 | ||
NM_123045.4 | NP_198503.3 | 17 - 18 | 2001 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |