Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344150.1 | NP_001329981.1 | 5 - 6 | 615 | 128 | |
NM_001344151.1 | NP_001329982.1 | 5 - 6 | 615 | 128 | ||
NM_001344152.1 | NP_001329979.1 | 4 - 5 | 686 | 128 | ||
NM_001344153.1 | NP_001329980.1 | 4 - 5 | 535 | 128 | ||
NM_123045.4 | NP_198503.3 | 4 - 5 | 535 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |