Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123053.5 | NP_568543.1 | 7 - 8 | 1533 | 146 | |
GenBank mRNA | AY069888.1 | - | 7 - 8 | 1521 | 146 | |
BT015174.1 | - | 7 - 8 | 1510 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |