Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085203.1 | NP_001078672.1 | 2 - 3 | 273 | 102 | |
NM_001344162.1 | NP_001331077.1 | 2 - 3 | 695 | 102 | ||
NM_123060.3 | NP_198518.1 | 2 - 3 | 848 | 102 | ||
GenBank mRNA | AF042196.1 | - | 2 - 3 | 273 | 102 | |
AY669790.1 | - | 2 - 3 | 150 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |