Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085203.1 | NP_001078672.1 | 8 - 9 | 931 | 125 | |
NM_001344162.1 | NP_001331077.1 | 7 - 8 | 1459 | 125 | ||
NM_123060.3 | NP_198518.1 | 8 - 9 | 1506 | 125 | ||
GenBank mRNA | AF042196.1 | - | 8 - 9 | 931 | 125 | |
AY669790.1 | - | 8 - 9 | 808 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |