Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344166.1 | NP_001318683.1 | 4 - 5 | 768 | 74 | |
NM_123077.3 | NP_198534.3 | 2 - 3 | 348 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |