Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344179.1 | NP_001331810.1 | 4 - 5 | 1324 | 140 | |
NM_001344180.1 | NP_001331809.1 | 5 - 6 | 1232 | 140 | ||
NM_123089.3 | NP_568547.1 | 5 - 6 | 1162 | 140 | ||
GenBank mRNA | AY056316.1 | - | 5 - 6 | 1153 | 140 | |
BT015776.1 | - | 5 - 6 | 1144 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |