Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344207.1 | NP_001332118.1 | 1 - 2 | 540 | 110 | |
NM_001344209.1 | NP_001332119.1 | 1 - 2 | 540 | 110 | ||
NM_123109.5 | NP_198566.2 | 1 - 2 | 540 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |