Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARLA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344213.1 | NP_001331082.1 | 1 - 2 | 249 | 75 | |
NM_123127.4 | NP_568553.1 | 3 - 4 | 515 | 75 | ||
GenBank mRNA | AY062658.1 | - | 3 - 4 | 400 | 75 | |
AY085271.1 | - | 3 - 4 | 399 | 75 | ||
AY114643.1 | - | 3 - 4 | 270 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |