Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARLA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344213.1 | NP_001331082.1 | 2 - 3 | 411 | 108 | |
NM_123127.4 | NP_568553.1 | 4 - 5 | 677 | 108 | ||
GenBank mRNA | AY062658.1 | - | 4 - 5 | 562 | 108 | |
AY085271.1 | - | 4 - 5 | 561 | 108 | ||
AY114643.1 | - | 4 - 5 | 432 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |