Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337489.1 | NP_001325638.1 | 2 - 3 | 770 | 133 | |
NM_111252.5 | NP_187031.2 | 3 - 4 | 682 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |