Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337509.1 | NP_001326045.1 | 10 - 11 | 1205 | 78 | |
NM_111295.4 | NP_187074.1 | 16 - 17 | 1805 | 78 | ||
GenBank mRNA | AF441079.1 | - | 16 - 17 | 1723 | 78 | |
AK226884.1 | - | 16 - 17 | 1748 | 78 | ||
AY090938.1 | - | 16 - 17 | 1744 | 78 | ||
AY117340.1 | - | 16 - 17 | 1594 | 78 | ||
KJ138841.1 | - | 16 - 17 | 1594 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |