Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111295.4 | NP_187074.1 | 4 - 5 | 655 | 68 | |
GenBank mRNA | AF441079.1 | - | 4 - 5 | 573 | 68 | |
AK226884.1 | - | 4 - 5 | 598 | 68 | ||
AY090938.1 | - | 4 - 5 | 594 | 68 | ||
AY117340.1 | - | 4 - 5 | 444 | 68 | ||
KJ138841.1 | - | 4 - 5 | 444 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |