Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111297.4 | NP_187076.2 | 1 - 2 | 450 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |