Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111297.4 | NP_187076.2 | 9 - 10 | 1838 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |