Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114131.3 | NP_189850.1 | 10 - 11 | 2792 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |