Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114228.3 | NP_189946.1 | 2 - 3 | 2131 | 64 | |
GenBank mRNA | AB005805.1 | - | 2 - 3 | 2092 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |