Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339113.1 | NP_001327805.1 | 4 - 5 | 947 | 96 | |
NM_114254.3 | NP_189972.2 | 4 - 5 | 947 | 96 | ||
GenBank mRNA | AY072099.1 | - | 4 - 5 | 907 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |