Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161190.2 | NP_001154662.2 | 15 - 16 | 2694 | 89 | |
NM_001161191.3 | NP_001154663.2 | 15 - 16 | 2812 | 89 | ||
NM_001339114.1 | NP_001325812.1 | 15 - 16 | 2822 | 89 | ||
NM_001339115.1 | NP_001325811.1 | 15 - 16 | 2825 | 89 | ||
NM_001339116.1 | NP_001325813.1 | 10 - 11 | 2195 | 89 | ||
NM_114260.1 | NP_189978.1 | 13 - 14 | 2547 | 89 | ||
GenBank mRNA | DQ479972.1 | - | 13 - 14 | 2547 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |