Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339152.1 | NP_001319682.1 | 8 - 9 | 1870 | 78 | |
NM_001339153.1 | NP_001327210.1 | 8 - 9 | 2520 | 78 | ||
NM_114322.5 | NP_190040.3 | 9 - 10 | 1980 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |