Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114328.4 | NP_190046.2 | 2 - 3 | 378 | 85 | |
GenBank mRNA | AY062653.1 | - | 2 - 3 | 336 | 85 | |
BT008382.1 | - | 2 - 3 | 275 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |