Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114370.4 | NP_190087.1 | 1 - 2 | 526 | 100 | |
GenBank mRNA | AY050427.1 | - | 1 - 2 | 380 | 100 | |
AY093993.1 | - | 1 - 2 | 342 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |