Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339194.1 | NP_001327790.1 | 15 - 16 | 2224 | 66 | |
NM_001339195.1 | NP_001327789.1 | 14 - 15 | 2131 | 66 | ||
NM_001339196.1 | NP_001327788.1 | 14 - 15 | 2331 | 66 | ||
NM_001339197.1 | NP_001327787.1 | 15 - 16 | 2224 | 66 | ||
NM_114382.5 | NP_190099.3 | 14 - 15 | 2112 | 66 | ||
GenBank mRNA | AB247155.1 | - | 14 - 15 | 2072 | 66 | |
DQ508794.2 | - | 14 - 15 | 2072 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |