Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339194.1 | NP_001327790.1 | 4 - 5 | 625 | 77 | |
NM_001339195.1 | NP_001327789.1 | 3 - 4 | 532 | 77 | ||
NM_001339196.1 | NP_001327788.1 | 3 - 4 | 732 | 77 | ||
NM_001339197.1 | NP_001327787.1 | 4 - 5 | 625 | 77 | ||
NM_114382.5 | NP_190099.3 | 3 - 4 | 513 | 77 | ||
GenBank mRNA | AB247155.1 | - | 3 - 4 | 473 | 77 | |
DQ508794.2 | - | 3 - 4 | 473 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |