Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114398.2 | NP_566879.1 | 11 - 12 | 1290 | 85 | |
GenBank mRNA | AF369564.1 | - | 11 - 12 | 1144 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |