Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126612.3 | NP_565331.1 | 11 - 12 | 1222 | 102 | |
NM_179609.1 | NP_849940.1 | 11 - 12 | 1212 | 102 | ||
GenBank mRNA | AY056192.1 | - | 11 - 12 | 1183 | 102 | |
AY113962.1 | - | 10 - 11 | 1025 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |