Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126612.3 | NP_565331.1 | 5 - 6 | 779 | 123 | |
NM_179609.1 | NP_849940.1 | 5 - 6 | 769 | 123 | ||
GenBank mRNA | AY056192.1 | - | 5 - 6 | 740 | 123 | |
AY113962.1 | - | 4 - 5 | 582 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |