Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124823.2 | NP_001118295.1 | 2 - 3 | 392 | 111 | |
NM_001335365.1 | NP_001324917.1 | 2 - 3 | 286 | 111 | ||
NM_126831.4 | NP_178872.2 | 2 - 3 | 404 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |