Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126855.4 | NP_565351.1 | 2 - 3 | 645 | 130 | |
GenBank mRNA | AF367339.1 | - | 2 - 3 | 530 | 130 | |
AY093990.1 | - | 1 - 2 | 302 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |