Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036274.1 | NP_001031351.1 | 8 - 9 | 1079 | 67 | |
NM_001335380.1 | NP_001323740.1 | 8 - 9 | 1280 | 67 | ||
NM_001335381.1 | NP_001323739.1 | 8 - 9 | 1208 | 67 | ||
NM_126893.4 | NP_178937.2 | 8 - 9 | 1079 | 67 | ||
GenBank mRNA | BT004306.1 | - | 8 - 9 | 1079 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |