Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GONST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202595.1 | NP_001189524.1 | 8 - 9 | 1085 | 71 | |
NM_001335395.1 | NP_001318219.1 | 6 - 7 | 1371 | 71 | ||
NM_001335396.1 | NP_001324974.1 | 6 - 7 | 1536 | 71 | ||
NM_001335397.1 | NP_001324975.1 | 5 - 6 | 878 | 71 | ||
NM_001335398.1 | NP_001324973.1 | 7 - 8 | 1144 | 71 | ||
NM_126944.5 | NP_565357.1 | 7 - 8 | 1209 | 71 | ||
GenBank mRNA | KR265320.1 | - | 6 - 7 | 954 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |