Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335399.1 | NP_001318220.1 | 2 - 3 | 189 | 110 | |
NM_001335400.1 | NP_001324219.1 | 1 - 2 | 259 | 110 | ||
NM_126956.4 | NP_179000.3 | 1 - 2 | 259 | 110 | ||
GenBank mRNA | AY094412.1 | - | 1 - 2 | 75 | 110 | |
FJ708692.1 | - | 1 - 2 | 86 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |