Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161044.1 | NP_001154516.1 | 7 - 8 | 1115 | 102 | |
NM_001335415.1 | NP_001323471.1 | 7 - 8 | 1144 | 102 | ||
NM_126984.4 | NP_565362.1 | 7 - 8 | 1270 | 102 | ||
NM_126985.6 | NP_565363.2 | 7 - 8 | 1115 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |