Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084418.1 | NP_001077887.1 | 2 - 3 | 208 | 145 | |
NM_001084419.1 | NP_001077888.1 | 3 - 4 | 408 | 145 | ||
NM_126989.2 | NP_179032.1 | 3 - 4 | 408 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |