Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084418.1 | NP_001077887.1 | 6 - 7 | 673 | 93 | |
NM_001084419.1 | NP_001077888.1 | 7 - 8 | 873 | 93 | ||
NM_126989.2 | NP_179032.1 | 7 - 8 | 873 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |