Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100149.4 | NP_171769.1 | 9 - 10 | 1489 | 101 | |
NM_202014.1 | NP_973743.1 | 9 - 10 | 1366 | 101 | ||
GenBank mRNA | AY070068.1 | - | 9 - 10 | 1363 | 101 | |
AY096437.1 | - | 9 - 10 | 1226 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |