Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123745.2 | NP_001117217.1 | 10 - 11 | 1466 | 98 | |
NM_100166.4 | NP_563666.1 | 11 - 12 | 1316 | 98 | ||
NM_179247.6 | NP_849578.5 | 11 - 12 | 1397 | 98 | ||
NM_202017.3 | NP_973746.3 | 10 - 11 | 1335 | 98 | ||
GenBank mRNA | AY049274.1 | - | 11 - 12 | 1177 | 98 | |
AY062763.1 | - | 11 - 12 | 1178 | 98 | ||
BT001137.1 | - | 11 - 12 | 1121 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |