Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035893.1 | NP_001030970.1 | 1 - 2 | 134 | 73 | |
NM_100396.4 | NP_172010.1 | 1 - 2 | 263 | 73 | ||
NM_202032.2 | NP_973761.1 | 1 - 2 | 134 | 73 | ||
GenBank mRNA | AY050379.1 | - | 1 - 2 | 123 | 73 | |
L13922.1 | - | 1 - 2 | 102 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |